missing translation for 'onlineSavingsMsg'
Learn More

Hydrazine Hydrate Solution, 20 to 24%, Honeywell Fluka™

Product Code. 15675810 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Poly bottle
Unit Size:
1 L
250mL
Product Code. Quantity Packaging unitSize
15675810 1 L Poly bottle 1 L
15685810 1 L Poly bottle 250mL
2 options
This item is not returnable. View return policy

Product Code. 15675810

Brand: Honeywell-Fluka 538471L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss. p.a., 24-26% in H2O (RT)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10217-52-4
Molecular Formula H4N2
Molecular Weight (g/mol) 32.05
MDL Number MFCD00149931
InChI Key OAKJQQAXSVQMHS-UHFFFAOYSA-N
Synonym hydrazine hydrate, hydrazine monohydrate, hydrazinium hydroxide, hydrazine hydroxide, hydrazine, monohydrate, hydrazine, hydrate, idrazina idrata italian, hydrazinemonohydrate, unii-kyd297831p, ccris 7739
PubChem CID 24654
ChEBI CHEBI:35511
SMILES NN

Specifications

Quantity 1 L
Linear Formula NH2NH2 · H2O
UN Number UN3293
Beilstein 878137
IUPAC Name hydrazine
Formula Weight 50.06g/mol
Packaging Poly bottle
Appearance Colorless
Density 1.01g/mL at 20°C
Chemical Name or Material Hydrazine hydrate solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.