missing translation for 'onlineSavingsMsg'
Learn More

Hydrazine monohydrate, 98+%

Product Code. 11498880
Change view
Click to view available options
Quantity:
50 g
250 g
500 g
2500 g
Unit Size:
2500g
250g
500g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11498880 250 g 250g
11488880 50 g 50g
11447866 500 g 500g
11457866 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11498880 Supplier Thermo Scientific Alfa Aesar Supplier No. A14005.30
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7803-57-8 | H6N2O | 50.06 g/mol

Hydrazine monohydrate is used as a reducing agent. It is also used in water treatment, especially effluents and industrial boilers, metal treatment and mine extraction process, and synthesis of active ingredients in the pharmaceutical industry like 3-amino-1,2,4-triazole cefazolin, rizatriptan, anastrozole, fluconazole, metazachlor metamitron pyrazole, metribuzin, paclobutrazol, diclobutrazole, propiconazole, and triadimefon. It is involved in the preparation of heterocycles like pyrazoles and pyridazines. It is actively involved in the Wolff-Kishner reduction, which converts the carbonyl group of a ketone into a methylene bridge through a hydrazone intermediate. Further, it is used to cleave N-alkylated phthalimide derivatives. In the Einhorn-Brunner reaction, it reacts with imides to get triazoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hydrazine monohydrate is used as a reducing agent. It is also used in water treatment, especially effluents and industrial boilers, metal treatment and mine extraction process, and synthesis of active ingredients in the pharmaceutical industry like 3-amino-1,2,4-triazole cefazolin, rizatriptan, anastrozole, fluconazole, metazachlor metamitron pyrazole, metribuzin, paclobutrazol, diclobutrazole, propiconazole, and triadimefon. It is involved in the preparation of heterocycles like pyrazoles and pyridazines. It is actively involved in the Wolff-Kishner reduction, which converts the carbonyl group of a ketone into a methylene bridge through a hydrazone intermediate. Further, it is used to cleave N-alkylated phthalimide derivatives. In the Einhorn-Brunner reaction, it reacts with imides to get triazoles.

Solubility
Miscible with water and alcohol. Immiscible with chloroform, hydrocarbons and ether.

Notes
Special handling precautions required. View MSDS prior to purchase. MSDS are available online at www.alfa.comIncompatible with oxidizing agents, oxygen, copper, organic materials and zinc.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7803-57-8
Molecular Formula H6N2O
Molecular Weight (g/mol) 50.06
MDL Number MFCD00149931
InChI Key IKDUDTNKRLTJSI-UHFFFAOYSA-N
Synonym hydrazine hydrate, hydrazine monohydrate, hydrazinium hydroxide, hydrazine hydroxide, hydrazine, monohydrate, hydrazine, hydrate, idrazina idrata italian, hydrazinemonohydrate, unii-kyd297831p, ccris 7739
PubChem CID 24654
ChEBI CHEBI:35511
SMILES O.NN

Specifications

Boiling Point 120°C to 121°C
Melting Point -52°C
Quantity 250 g
Linear Formula H2NNH2·H2O
UN Number UN2030
Flash Point 75°C (167°F)
Merck Index 14,4771
Solubility Information Miscible with water and alcohol. Immiscible with chloroform,hydrocarbons and ether.
IUPAC Name hydrazine;hydrate
Formula Weight 50.06 (32.05 Anhydrous)
Percent Purity ≥98%
Odor Amine-like
Refractive Index 1.431
Density 1.027
Chemical Name or Material Hydrazine monohydrate
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hydrazine monohydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H350-May cause cancer.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: hydrazine monohydrate, hydrazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.