missing translation for 'onlineSavingsMsg'
Learn More

Hydrazine monohydrochloride, 98+%

Product Code. 10164120 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic Drum
Unit Size:
1kg
250g
5kg
Product Code. Quantity Packaging unitSize
10164120 250 g Glass bottle 250g
10266350 1 kg Glass bottle 1kg
10668314 5 kg Plastic Drum 5kg
3 options
This item is not returnable. View return policy

Product Code. 10164120

Brand: Thermo Scientific Acros 201992500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2644-70-4 | H4N2·HCl | 68.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2644-70-4
Molecular Formula H4N2·HCl
Molecular Weight (g/mol) 68.51
MDL Number MFCD00044368
InChI Key BIVUUOPIAYRCAP-UHFFFAOYSA-N
Synonym hydrazine hydrochloride, hydrazine monohydrochloride, hydrazine, monohydrochloride, hydrazine,chloride, diamine hydrochloride, hydrazine.hydrochloride, hydrazine,hydrochloride, hydrazine mono hcl, nh2nh2hcl, pubchem17422
PubChem CID 115031
IUPAC Name hydrazine;hydrochloride
SMILES NN.Cl

Specifications

Melting Point 89°C to 93°C
Color White
Physical Form Powder
Quantity 250 g
Assay Percent Range 98% min. (Argentometry)
Linear Formula H2NNH2·HCl
Solubility Information Solubility in water: 370g/L (20°C). Other solubilities: slightly soluble in alcohol,insoluble in most organic solvents,soluble in liquid ammoniak
Formula Weight 68.51
Percent Purity ≥98%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Hydrazine monohydrochloride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hydrazine monohydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H350-May cause cancer.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.