missing translation for 'onlineSavingsMsg'
Learn More

Hydrazine Sulfate-15N2, TRC

Product Code. 30133477
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133477 50 mg 50mg
30133318 500 mg 500mg
30009373 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133477 Supplier Toronto Research Chemicals Supplier No. TRCH71800250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88491-70-7
Molecular Formula H2 O4 S . H4 15N2
Molecular Weight (g/mol) 132.11
Synonym Hydrazine-1,2-15N2, sulfate (1:1), Hydrazine-15N2, sulfate (1:1) (9CI)
SMILES [15NH2][15NH2].OS(=O)(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/H4N2.H2O4S/c1-2;1-5(2,3)4/h1-2H2;(H2,1,2,3,4)/i1+1,2+1;
Formula Weight 131.9989
Percent Purity >95%
Chemical Name or Material Hydrazine-15N2 sulfate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.