missing translation for 'onlineSavingsMsg'
Learn More

Hydrazine Sulfate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Product Code. 15557984 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
125 g
Packaging:
Poly Bottle
Unit Size:
125g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15557984 125 g Poly Bottle 125g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15557984 Supplier Avantor J.T.Baker Supplier No. 217704

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Specifications

CAS 10034-93-2
Packaging Poly Bottle
Molecular Formula H6N2O4S
Quantity 125 g
Specific Gravity 1L = 1.37kg
Synonym hydrazine sulfate, hydrazine monosulfate, hydrazine, sulfate, hydrazine sulphate, hydrazinium sulfate, hydrazonium sulfate, hydrazine sulfate 1:1, segidrin, sehydrin, diamine sulfate
InChI Key ZGCHATBSUIJLRL-UHFFFAOYSA-N
SMILES NN.OS(=O)(=O)O
IUPAC Name hydrazine;sulfuric acid
Molecular Weight (g/mol) 130.118
PubChem CID 24842
Formula Weight 130.12
Grade BAKER ANALYZED™ ACS Reagent
Physical Form Crystals
Chemical Name or Material Hydrazine Sulfate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.