missing translation for 'onlineSavingsMsg'
Learn More

Hydrazine Sulfate Salt, TRC

Product Code. 30133243
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133243 25 g 25g
18033820 5 g 5g
18080410 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133243 Supplier Toronto Research Chemicals Supplier No. TRCH71800025G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10034-93-2
Molecular Formula H2 O4 S . H4 N2
Molecular Weight (g/mol) 130.12
Synonym Hydrazine, sulfate (1:1), Hydrazine dihydrogen sulfate (salt), Hydrazine hydrogen sulfate, Hydrazine hydrosulfate, Hydrazine monosulfate, Hydrazine sulfate, Hydrazinium sulfate, Hydrazinium(2+) sulfate, Hydrazonium sulfate, NSC 150014, NSC 215190, Segidrin, Sehydrin
IUPAC Name hydrazine;sulfuric acid
SMILES NN.OS(=O)(=O)O

Specifications

Quantity 25 g
InChI Formula InChI=1S/H4N2.H2O4S/c1-2;1-5(2,3)4/h1-2H2;(H2,1,2,3,4)
Formula Weight 130.0048
Chemical Name or Material Hydrazine Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.