missing translation for 'onlineSavingsMsg'
Learn More

Hydrazinecarboxaldehyde, TRC

Product Code. 30404192
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30404192 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30404192 Supplier Toronto Research Chemicals Supplier No. TRCF69290510G
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hydrazinecarboxaldehyde
CAS 624-84-0
Molecular Formula CH4N2O
Quantity 10 g
Synonym Formylhydrazine, Carbazaldehyde, Formal Hydrazine, Formhydrazide, Formic Hydrazide, Formohydrazide, Formylhydrazide, Formylhydrazine, Monoformylhydrazine, N-Formylhydrazine, NSC 72391
InChI Formula InChI=1S/CH4N2O/c2-3-1-4/h1H,2H2,(H,3,4)
SMILES NNC=O
IUPAC Name formohydrazide
Molecular Weight (g/mol) 60.06
Formula Weight 60.0324
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.