missing translation for 'onlineSavingsMsg'
Learn More

Hydriodic acid, for analysis, 57 wt.% aqueous solution, distilled, stabilized

Product Code. 10441241 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mL
250 mL
1 L
Unit Size:
1 L
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10441241 50 mL 50mL
10103930 250 mL 250mL
11984101 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10441241 Supplier Thermo Scientific Acros Supplier No. 198370500
  • ONLINE EXCLUSIVE PRICE
    45.25 €
    41.80 € / 50mL
    Save 3.45 €
    7% Off
View all promotions
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10034-85-2 | HI | 127.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10034-85-2
Molecular Formula HI
Molecular Weight (g/mol) 127.91
MDL Number MFCD00011347
InChI Key XMBWDFGMSWQBCA-UHFFFAOYSA-N
IUPAC Name hydrogen iodide
SMILES I

Specifications

Boiling Point 127.0°C
Density 1.7000 g/mL
Packaging Glass bottle
Specific Gravity 1.7
Quantity 50 mL
Linear Formula HI
Fieser 01,449; 02,213; 06,281; 07,173; 08,246; 09,238; 10,199; 11,255; 15,166
Merck Index 15, 4814
Solubility Information Solubility aq. soln.: soluble.
Formula Weight 127.91
Grade Analysis
Chemical Name or Material Hydriodic acid
Show More Show Less
compliance-icons
Product Identifier
  • Hydriodic acid, aqueous solution
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Hydriodic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.