missing translation for 'onlineSavingsMsg'
Learn More

Hydriodic Acid, stabilized, Honeywell

Product Code. 15671910 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
50 mL
250 mL
Unit Size:
250mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15671910 50 mL 50mL
15661910 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15671910 Supplier Honeywell Fluka Supplier No. 21002150ML
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 10034-85-2
Molecular Formula HI
Molecular Weight (g/mol) 127.91
MDL Number MFCD00011347
InChI Key XMBWDFGMSWQBCA-UHFFFAOYSA-N
Synonym hydriodic acid, hydrogen iodide, hydroiodic acid, hydriotic acid, iodium, yodo, jodwasserstoff, iodine-125, unii-694c0eft9q, iodo group
PubChem CID 24841
ChEBI CHEBI:43451
IUPAC Name hydrogen iodide
SMILES I

Specifications

Density 1.701g/mL at 25°C
Quantity 50 mL
UN Number 1787
Formula Weight 127.91g/mol
Percent Purity 99.95%
Chemical Name or Material Hydriodic Acid, Distilled, Stabilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.