missing translation for 'onlineSavingsMsg'
Learn More

Hydrobromic Acid, 47.0 to 49.0%, ACS Reagent, Honeywell™ Fluka™

Product Code. 15675520 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Glass bottle
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15675520 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15675520 Supplier Honeywell-Fluka Supplier No. 438065500ML
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS Reagent, 48%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

pH Acidic
Color Yellow
CAS 10035-10-6
Melting Point ∼-11°C
Boiling Point 126°C (1013hPa)
Density 1.49g/mL at 25°C
Odor Stinging
Packaging Glass bottle
Vapor Pressure 320psi (21.1°C), 8mmHg (25°C)
Quantity 500 mL
Molecular Formula BrH
Physical Form Liquid
MDL Number MFCD00011323
UN Number UN1788
Beilstein 3587158
Synonym hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
Solubility Information Completely miscible in water
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-N
SMILES Br
IUPAC Name bromane
Molecular Weight (g/mol) 80.91
PubChem CID 260
ChEBI CHEBI:47266
Formula Weight 80.91g/mol
Percent Purity 47.0 to 49.0%
Chemical Name or Material Hydrobromic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.