missing translation for 'onlineSavingsMsg'
Learn More

Hydrobromic Acid (47-48% in aqueous solution), TRC

Product Code. 30406079
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30406079 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30406079 Supplier Toronto Research Chemicals Supplier No. TRCH714585100ML

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hydrobromic Acid
CAS 10035-10-6
Molecular Formula HBr
Quantity 100 mL
Synonym Bromohydric Acid, Hydrogen Bromide, Hydrogen Monobromide, NSC 606640
InChI Formula InChI=1S/BrH/h1H
SMILES Br
IUPAC Name bromane
Molecular Weight (g/mol) 80.91
Formula Weight 79.9262
Concentration or Composition (by Analyte or Components) 47-48 %
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.