missing translation for 'onlineSavingsMsg'
Learn More

Hydrobromic Acid, 47% to 49%, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Product Code. 15567784 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Narrow Mouth Amber Glass Bottle
Unit Size:
4 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15567784 500 mL Narrow Mouth Amber Glass Bottle 500mL
15577784 4 L Narrow Mouth Amber Glass Bottle 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15567784 Supplier Avantor J.T.Baker Supplier No. 016001
  • ONLINE EXCLUSIVE PRICE
    356.00 €
    329.01 € / 500mL
    Save 26.99 €
    7% Off
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

    For Laboratory,Research,or Manufacturing Use

Chemical Identifiers

CAS 10035-10-6
Molecular Formula BrH
Molecular Weight (g/mol) 80.91
MDL Number MFCD00011323
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-N
Synonym hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
PubChem CID 260
ChEBI CHEBI:47266
SMILES Br

Specifications

Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.50kg
Quantity 500 mL
IUPAC Name hydrogen bromide
Formula Weight 80.92
Percent Purity 47 to 49%
Grade Meets ACS reagent chemical requirements
Chemical Name or Material Hydrobromic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.