missing translation for 'onlineSavingsMsg'
Learn More

Hydrobromic acid, ACS reagent, 48%, Solstice

Product Code. 16695773
Change view
Click to view available options
Quantity:
10 L
Unit Size:
10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16695773 10 L 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16695773 Supplier Fluka Supplier No. 24426010L
  • ONLINE EXCLUSIVE PRICE
    1555.00 €
    1405.65 € / 10 L
    Save 149.35 €
    9% Off
 Restricted Item
Estimated Shipment: 02-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Color Light Yellow
CAS 7732-18-5
Melting Point -11°C
Boiling Point 126°C (1,013 hPa)
CAS Min % ≥ 50%
CAS Max % <70%
Density 1.490 g/cm3 (20°C)
Quantity 10 L
Molecular Formula H2O
Linear Formula HBr
Beilstein 3587158
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILES O
IUPAC Name water
Molecular Weight (g/mol) 18.02
Grade ACS
Chemical Name or Material Hydrobromic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.