missing translation for 'onlineSavingsMsg'
Learn More

Hydrobromic acid, ACS reagent, ca. 48% solution in water

Product Code. 10761434 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
2.5 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10761434 2.5 L Glass bottle 2.5 L
10025870 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10761434 Supplier Thermo Scientific Acros Supplier No. 423780025
  • ONLINE EXCLUSIVE PRICE
    286.00 €
    277.00 € / 2.5 L
    Save 9.00 €
    3% Off
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hydrobromic acid, 47.0 to 49.0%, BrH, CAS Number-10035-10-6, 7732-18-5, acide bromhydrique, acido bromidrico, anhydrous hydrobromic acid, bromowodor, bromowodor polish, bromwasserstoff, broomwaterstof, hydrobromic acid, hydrobromide, hydrogen bromide, 2.5L, 126.0 deg.C to 128.0 deg.C, CHEBI:47266 | CAS: 10035-10-6 | BrH | 80.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10035-10-6 , 7732-18-5
Molecular Formula BrH
Molecular Weight (g/mol) 80.91
MDL Number MFCD00011323
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-N
Synonym hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
PubChem CID 260
ChEBI CHEBI:47266
IUPAC Name hydrogen bromide
SMILES Br

Specifications

Color Colorless to Yellow
Melting Point -11.0°C
Boiling Point 126.0°C to 128.0°C
Density 1.4800g/mL
Heavy Metals (as Pb) 5ppm max.
Packaging Glass bottle
Residue after Ignition 0.002% max.
Specific Gravity 1.48
Quantity 2.5 L
Linear Formula HBr
Physical Form Liquid
Fieser 01,450; 02,214; 03,154; 04,249; 05,332; 06,282; 10,200; 15,273
Merck Index 15, 4816
Solubility Information Solubility in water: soluble.
Formula Weight 80.9
Percent Purity 47.0 to 49.0%
Grade ACS Reagent
Chemical Name or Material Hydrobromic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Hydrobromic acid, ca. 48 wt% solution in water
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.