Learn More
Description
For siliconizing glass surfaces use Thermo Scientific™ Hydrocarbon Soluble Siliconizing Fluid. This short chain, clear polymeric silicone fluid consists primarily of dichlorooctamethyltetrasiloxane and is soluble in organic solvents. When applied to glass, quartz or similar products, the unhydrolyzed chlorines present on the chain react with surface silanols to form a neutral, hydrophobic and tiightly bonded film over the entire surface. It is excellent for modifying metals, certain plastics, ceramics and fiber optics.
Hydrocarbon Soluble Siliconizing Fluid is acidic and care should be taken to avoid corrosion of metal that comes into contact with the liquid.
The fluid is acidic only during application; after application, the surface is neutral.
Specifications
Specifications
| For Use With | GC |
| Hazards | Corrosive to Eyes/Skin/Respiratory Tract |
| Standard Components | 1,7-Dichloro-1,1,3,3,5,5,7,7-Octamethyltetrasiloxane |
| Quantity | 120 mL |
| Product Type | Hydrocarbon Soluble Siliconizing Fluid |
Safety and Handling
- Surfasil, 120 mL.
- Danger
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P310-Immediately call a POISON CENTER/doctor/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P501-Dispose of contents/container to .
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.