missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid Solution, 0.1M, Chem-Lab

Product Code. 11933113
Change view
Click to view available options
Quantity:
1 L
Packaging:
Polyethylene
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11933113 1 L Polyethylene 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11933113 Supplier Chem-Lab Supplier No. CL05.0307.1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used as a laboratory reagent and acidifying agent. Hydrochloric Acid Solution, 0.1M, Chem-Lab contains 3.646g HCl per liter of water. It is used for pH control, in the production of organic and inorganic compounds, and in the regeneration of ion exchange resins.

Hydrochloric acid is a corrosive, strong mineral acid with many industrial uses. It is a colorless, highly pungent solution of hydrogen chloride (HCl) in water. It is frequently used in pH buffer solutions, in the regeneration of ion exchange resins and in the production of chlorides and the synthesis of organic products.

Specifications

Color Colorless
CAS 7647-01-0, 7732-18-5
Composition 3.646g HCl per 1L H2O
Concentration 0.1mol/L
Density 1.00g/mL
Packaging Polyethylene
Normality 0.1N (±0.0002, 20°C)
Quantity 1 L
Molecular Formula ClH
Physical Form Liquid
MDL Number MFCD00011324 MFCD00792839
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES Cl
IUPAC Name hydrogen chloride
Molecular Weight (g/mol) 36.46
PubChem CID 313
ChEBI CHEBI:17883
Formula Weight 36.46g/mol
Show More Show Less
compliance-icons
Product Identifier
  • Hydrochloric acid 0.1 mol/l
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P390-Absorb spillage to prevent material damage.
Supplemental information
  • MIXTURE LIST-Contain: HCl/H2O
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.