missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid, 0.5N Standardized Solution

Product Code. 11323777
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
Unit Size:
1 L
20 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11323777 4 L 4 L
11333777 1 L 1 L
11313777 20 L 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11323777 Supplier Thermo Scientific Alfa Aesar Supplier No. 035642.K7
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7647-01-0 | ClH | 36.46 g/mol

Hydrochloric acid, 0.5N standardized solution is used for volumetric analysis in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0 , 7732-18-5
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name chlorane
SMILES Cl

Tekniske data

Appearance Colorless liquid
Color Colorless
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Quantity 4 L
Physical Form Liquid
Solubility Information Miscible with water at room temperature. Also miscible with methanol,ethanol and ether.
Formula Weight 36.46
Concentration or Composition (by Analyte or Components) Hydrochloric acid: 4.13%; Water: 95.87%
Chemical Name or Material Hydrochloric acid
compliance-icons
Produktidentifikator
  • Hydrochloric acid, 0.5N Standardized Solution
Signalord
  • Warning
Farekategori
  • Substances and mixtures corrosive to metals Category 1
Faresætning
  • H290-May be corrosive to metals.
Sikkerhedssætning
  • P234-Keep only in original container.
  • P390-Absorb spillage to prevent material damage.
Supplerende oplysninger
  • MIXTURE LIST-Contain: Hydrochloric acid

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.