missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid 1 M For Laboratory Use and Titrations, Chem Lab

Product Code. 11923133
Change view
Click to view available options
Hydrochloric Acid 1 M For Laboratory Use and Titrations, 25 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11923133 Hydrochloric Acid 1 M For Laboratory Use and Titrations, 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11923133 Supplier Chem-Lab Supplier No. CL05.0311.9025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Color Colorless
CAS 7647-01-0
Melting Point 0°C
Boiling Point 100°C
Concentration 1 M
Density 1.02 g/mL
Packaging Polyethylene
Traceability to NIST Reference standards acc. NIST SRM 84l
Quantity 25 L
Physical Form Liquid
Molecular Weight (g/mol) 36.46
Show More Show Less
compliance-icons
Product Identifier
  • Hydrochloric acid 1 mol/l
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P390-Absorb spillage to prevent material damage.
Supplemental information
  • MIXTURE LIST-Contain: HCl/H2O
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.