missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid 10% w/v, Reagecon

Product Code. 13175509 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13175509 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13175509 Supplier Reagecon Supplier No. H10WV1
  • ONLINE EXCLUSIVE PRICE
    134.00 €
    126.02 € / 1 L
    Save 7.98 €
    5% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hydrochloric Acid 10% w/v

  • Hydrochloric Acid 10% w/v

Specifications

Concentration 10% w/v
Packaging HDPE Bottle GL45
Quantity 1 L
Physical Form Liquid
compliance-icons
Product Identifier
  • HYDROCHLORIC ACID 10% w/v
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
Supplemental information
  • MIXTURE LIST-Contain: hydrochloric acid 37%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.