missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 1N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T. Baker™

Product Code. 15578334 Shop All J.T.Baker Products
Click to view available options
Quantity:
1 L
Packaging:
Narrow Mouth Poly Bottle
Unit Size:
1 L
This item is not returnable. View return policy

Product Code. 15578334

Brand: Avantor J.T.Baker 562002

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
SMILES Cl

Specifications

Packaging Narrow Mouth Poly Bottle
Specific Gravity 1L = 1.00kg
Quantity 1 L
IUPAC Name chlorane
Formula Weight 36.46
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material Hydrochloric Acid, 1N Volumetric Solution
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
  • P390-Absorb spillage to prevent material damage.
  • P406-Store in corrosive resistant/ container with a resistant inner liner.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.