missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, 25%, BAKER ANALYZED, J.T. Baker™
Shop All J.T.Baker Products
Click to view available options
Quantity:
2.5 L
Packaging:
Polyethylene (HDPE)
Unit Size:
2.5 L
Description
- Label provides an actual Lot Analysis as a guarantee of purity and consistency
- Analytical methods used for these determinations are based on standard test procedures described in technical publications
- Improvements developed by J.T. Baker are employed to achieve a higher degree of reliability
Specifications
Specifications
| Color | Colorless |
| CAS | 7647-01-0 |
| Melting Point | 0°C |
| Boiling Point | 85°C |
| Density | 1.0g/cm3 at 20°C |
| Packaging | Polyethylene (HDPE) |
| Residue after Ignition | Max. 5 ppm |
| Quantity | 2.5 L |
| Molecular Formula | ClH |
| Physical Form | Liquid |
| Show More |
Safety and Handling
Signal Word
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction