missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric acid, meets analytical spec. of BP, NF, Ph. Eur., fuming, 36.5-38%, Honeywell Fluka™
Shop All Honeywell Fluka Products

Click to view available options
Quantity:
1 L
2.5 L
30 kg
4 x 2.5 L
6 x 1 L
Packaging:
Glass Bottle
Paper / Poly Bag
Plastic Bottle
Unit Size:
1 L
2.5 L
30kg
Pack of 4
Pack of 6
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Color | Yellow |
| Concentration | 36.5 to 38% |
| Density | 1.2g/mL at 25°C |
| Packaging | Plastic Bottle |
| Residue after Ignition | ≤0.002% (as SO4) |
| Vapor Pressure | 3.23psi (21.1°C), 7.93psi (37.7°C) |
| Quantity | 1 L |
| UN Number | 1789 |
| Beilstein | 1098214 |
| IUPAC Name | chlorane |
| Show More |
Safety and Handling
Product Identifier
- Hydrochloric acid
- Danger
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity Category 3
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction