missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, ACS Reagent, 37%, Honeywell Fluka™

Product Code. 15642790 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
2.5 L
6 x 500 mL
4 x 4 L
189 L
Density:
1.2 g/mL at 25°C
Unit Size:
100mL
189 L
2.5 L
25mL
500mL
Pack of 4
Pack of 6
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Density unitSize
15642790 500 mL 1.2 g/mL at 25°C 500mL
15638350 25 mL 1.2 g/mL at 25°C 25mL
15602790 100 mL 1.2 g/mL at 25°C 100mL
15622790 2.5 L 1.2 g/mL at 25°C 2.5 L
15652790 6 x 500 mL 1.2 g/mL at 25°C Pack of 6
15662790 6 x 500 mL 1.2 g/mL at 25°C Pack of 6
15632790 4 x 4 L 1.2 g/mL at 25°C Pack of 4
15612790 189 L 1.2 g/mL at 25°C 189 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 15642790 Supplier Honeywell-Fluka Supplier No. 258148500ML
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ACS Reagent, 37%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
SMILES Cl

Specifications

Concentration 37%
Density 1.2 g/mL at 25°C
Residue after Ignition ≤5ppm
Vapor Pressure 3.23psi (21.1°C), 7.93psi (37.7°C)
Quantity 500 mL
UN Number 1789
Beilstein 1098214
IUPAC Name hydrogen chloride
Formula Weight 36.46g/mol
Grade ACS Reagent
Chemical Name or Material Hydrochloric Acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Hydrochloric acid
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.