missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid, for analysis, 25% solution in water

Product Code. 10269623 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Plastic bottle
Dimensione della confezione:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity Packaging Dimensione della confezione
10269623 1 L Plastic bottle 1 L
10432076 2.5 L Plastic bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 10269623 Fornitore Thermo Scientific Acros N. del fornitore 387800010
  • ONLINE EXCLUSIVE PRICE
    35.85 €
    33.10 € / 1 L
    Save 2.75 €
    7% Off
Visualizza tutte le promozioni
 Restricted Item
Estimated Shipment: 26-11-2026
Aggiungi al carrello
Aggiungi al carrello
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 7647-01-0 | ClH | 36.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 7647-01-0 , 7732-18-5
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifica

Color Colorless
Melting Point -70.0°C
Boiling Point 107.0°C
Density 1.1200g/mL
Packaging Plastic bottle
Residue after Ignition 0.0005% max.
Specific Gravity 1.12
Linear Formula HCl
Quantity 1 L
Physical Form Liquid
Merck Index 15, 4818
Solubility Information Solubility in water: soluble.
Formula Weight 36.45
Percent Purity ≥25.0%
Grade Analytical
Chemical Name or Material Hydrochloric acid
Vedi altri risultati Mostra meno risultati
compliance-icons compliance-icons
Identificativo prodotto
  • Hydrochloric acid, 25%
Avvertenza
  • Danger
Categoria di pericolo
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Indicazioni di pericolo
  • H290-May be corrosive to metals.
  • H335-May cause respiratory irritation.
  • H314-Causes severe skin burns and eye damage.
Consigli di prudenza
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Informazioni supplementari
  • MIXTURE LIST-Contain: Hydrochloric acid

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.