missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, Certified ACS Plus, 36.5 to 38.0%, Fisher Chemical™
Specifications
Specifications
| pH | <1 |
| Concentration | 36.5 to 38.0% (w/w) |
| Density | 1.27 (Air=1.0) |
| Heavy Metals (as Pb) | 0.1ppm max. |
| Identification | Pass Test |
| Packaging | Glass Bottle |
| Residue after Ignition | 4ppm max. |
| Specific Gravity | 1.18 |
| Vapor Pressure | 125mbar at 20°C |
| Quantity | 2.5 L |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?