missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., fuming, ≥37%, APHA: ≤10, Solstice

Product Code. 15633100
Change view
Click to view available options
Quantity:
1 L
2.5 L
30 kg
4 x 2.5 L
6 x 1 L
235 kg
72 x 2.5 L
Packaging:
Glass Bottle
Metal Drum
Plastic Bottle
Unit Size:
1 L
2.5 L
235kg
30kg
Pack of 4
Pack of 6
Pack of 72
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15633100 1 L Glass Bottle 1 L
15623100 1 L Plastic Bottle 1 L
15653100 2.5 L Glass Bottle 2.5 L
15643100 2.5 L Plastic Bottle 2.5 L
15623110 6 x 1 L Glass Bottle Pack of 6
15603110 6 x 1 L Plastic Bottle Pack of 6
15693100 4 x 2.5 L Glass Bottle Pack of 4
15683100 4 x 2.5 L Plastic Bottle Pack of 4
15673100 30 kg Metal Drum 30kg
15633110 72 x 2.5 L - Pack of 72
15663100 235 kg - 235kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Product Code. 15633100 Supplier Honeywell-Fluka Supplier No. 307211LGL
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., fuming,≥37%, APHA: ≤10

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
SMILES Cl

Specifications

Density 1.2g/mL at 25°C
Packaging Glass Bottle
Residue after Ignition ≤0.0005% (as SO4)
Vapor Pressure 3.23psi (21.1°C), 7.93psi (37.7°C)
Quantity 1 L
UN Number 1789
Beilstein 1098214
IUPAC Name chlorane
Formula Weight 36.46g/mol
Percent Purity ≥37%
Grade ACS/puriss. p.a.
Chemical Name or Material Hydrochloric Acid, Reag. Ph. Eur. reag. ISO
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Hydrochloric acid
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.