missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid solution 0.05mol/L, Fisher Chemical™
Shop All Fisher Chemical Products

Click to view available options
Quantity:
1000 mL
Packaging:
Bottle
Unit Size:
1000mL
Specifications
Specifications
| CAS | 7647-01-0, 7732-18-5 |
| Concentration | 0.05M (0.05N) |
| Packaging | Bottle |
| Quantity | 1000 mL |
| Molecular Formula | ClH |
| MDL Number | MFCD00011324 MFCD00792839 |
| Synonym | hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid |
| InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
| SMILES | Cl |
| IUPAC Name | hydrogen chloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction