missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid Solution 0.5M (0.5N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™

Product Code. 10315710 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 Ampule
6 Ampules
Unit Size:
1 ampoule
6 ampoules
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10315710 6 Ampules 6 ampoules
10590721 1 Ampule 1 ampoule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10315710 Supplier Fisher Chemical Supplier No. J/4330C/90
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ClH, CAS Number-7647-01-0, spirits of salt, hydrogen chloride, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof, acide chlorhydrique, chlorohydric acid, muriatic acid, chlorwasserstoff, hydrochloric acid, 6AMP, CHEBI:17883, Colorless to Yellow, VEXZGXHMUGYJMC-UHFFFAOYSA-N

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
SMILES Cl

Specifications

pH lt; 1
Color Colorless to Yellow
Packaging Ampoule
Quantity 6 Ampules
Physical Form Liquid
IUPAC Name hydrogen chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.