missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid Solution 1M (1N), NIST Standard Solution, ready to use, for volumetric analysis, Fisher Chemical™

Product Code. 10488020 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
2.5 L
5 L
10 L
Packaging:
Bag in box
HDPE plastic bottle
Plastic container
Unit Size:
10 L
2.5 L
5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10488020 5 L Plastic container 5 L
10487830 2.5 L HDPE plastic bottle 2.5 L
10284480 10 L Bag in box 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10488020 Supplier Fisher Chemical Supplier No. J/4320/21
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7647-01-0 | Molecular Formula: ClH | Molecular Weight: 36.46 g/mol

Hydrochloric Acid Solution 1M (1N), NIST Standard Solution, ready to use, for volumetric analysis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
SMILES Cl

Specifications

Color Colorless
Boiling Point 100°C
Packaging Plastic container
Quantity 5 L
Physical Form Liquid
IUPAC Name hydrogen chloride
Formula Weight 36.46g/mol
compliance-icons
Product Identifier
  • Hydrochloric acid, 1M, (1N)
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
  • P390-Absorb spillage to prevent material damage.
Supplemental information
  • MIXTURE LIST-Contain: Hydrochloric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.