missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid Solution 2M (2N), NIST Standard Solution, ready to use, for volumetric analysis, Fisher Chemical™

Product Code. 10020210 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Bag in box
HDPE plastic bottle
Unit Size:
1 L
10 L
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10020210 2.5 L HDPE plastic bottle 2.5 L
10214440 1 L HDPE plastic bottle 1 L
11458962 10 L Bag in box 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10020210 Supplier Fisher Chemical Supplier No. J/4315/17
  • ONLINE EXCLUSIVE PRICE
    112.48 €
    89.20 € / 2.5 L
    Save 23.28 €
    20% Off
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
SMILES Cl

Specifications

Color Colorless
Boiling Point 100°C
Packaging HDPE plastic bottle
Quantity 2.5 L
Physical Form Liquid
IUPAC Name hydrogen chloride
Formula Weight 36.46g/mol
compliance-icons
Product Identifier
  • Hydrochloric acid, 2M, (2N)
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
  • P390-Absorb spillage to prevent material damage.
Supplemental information
  • MIXTURE LIST-Contain: Hydrochloric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.