missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid Solution 2M (2N), Pure, Bench Reagent, Fisher Chemical

Product Code. 10447450 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
HDPE plastic bottle
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10447450 2.5 L HDPE plastic bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10447450 Supplier Fisher Chemical Supplier No. J/4250/17
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

2.5L, HDPE plastic bottle

TRUSTED_SUSTAINABILITY

Specifications

pH 1
Color Colorless to Yellow
Melting Point 0°C
Boiling Point 100°C
Packaging HDPE plastic bottle
Vapor Pressure 14mmHg at 20°C
Quantity 2.5 L
Physical Form Liquid
MDL Number 11324
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.