missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid Solution, Volumetric, 2.0M HCl (2.0N), Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 mL
2 L
Packaging:
Poly bottle
Unit Size:
2 L
500mL
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Specifications
| Color | Colorless |
| Density | 1.04 g/cm3 |
| Packaging | Poly bottle |
| Vapor Pressure | 190mmHg (25°C) |
| Quantity | 500 mL |
| Physical Form | Liquid |
| UN Number | UN1789 |
| Formula Weight | 36.46g/mol |
| Chemical Name or Material | Hydrochloric acid solution, Volumetric, 2.0;M HCl (2.0N) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction