missing translation for 'onlineSavingsMsg'
Learn More

Hydrochlorothiazide Dimer, TRC

Product Code. 17974925
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17974925 50 mg 50mg
17963495 5 mg 5mg
17970675 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17974925 Supplier Toronto Research Chemicals Supplier No. TRCA62937550MG
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 402824-96-8
Molecular Formula C15 H16 Cl2 N6 O8 S4
Molecular Weight (g/mol) 607.49
Synonym N-[[7-(Aminosulfonyl)-6-chloro-2,3-dihydro-1,1-dioxido-4H-1,2,4-benzothiadiazin-4-yl]methyl]-6-chloro-3,4-dihydro-2H-1,2,4-benzothiadiazine-7-sulfonamide 1,1-Dioxide
IUPAC Name 6-chloro-4-[[(6-chloro-1,1-dioxo-3,4-dihydro-2H-1λ^{6},2,4-benzothiadiazin-7-yl)sulfonylamino]methyl]-1,1-dioxo-2,3-dihydro-1λ^{6},2,4-benzothiadiazine-7-sulfonamide
SMILES NS(=O)(=O)c1cc2c(cc1Cl)N(CNS(=O)(=O)c3cc4c(NCNS4(=O)=O)cc3Cl)CNS2(=O)=O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C15H16Cl2N6O8S4/c16-8-1-10-14(34(28,29)20-5-19-10)4-13(8)33(26,27)21-6-23-7-22-35(30,31)15-3-12(32(18,24)25)9(17)2-11(15)23/h1-4,19-22H,5-7H2,(H2,18,24,25)
Formula Weight 605.9289
Chemical Name or Material 6-Chloro-N-[(6-chloro-7-sulfamoyl-2,3-dihydro-4H-1,2,4-benzothiadiazin-4-yl 1,1-Dioxide)methyl]-3,4-dihydro-2H-1,2,4-benzothia-diazine-7-sulfonamide 1,1-Dioxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.