missing translation for 'onlineSavingsMsg'
Learn More

Hydrochlorothiazide, TRC

Product Code. 30010541
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
Unit Size:
10g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010541 25 g 25g
17971775 10 g 10g
17954585 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010541 Supplier Toronto Research Chemicals Supplier No. TRCH71456025G
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-93-5
Molecular Formula C7 H8 Cl N3 O4 S2
Molecular Weight (g/mol) 297.74
Synonym Hydrochlorothiazide, 2H-1,2,4-Benzothiadiazine-7-sulfonamide, 6-chloro-3,4-dihydro-, 1,1-dioxide, 3,4-Dihydrochlorothiazide, 6-Chloro-3,4-dihydro-2H-1,2,4-benzothiadiazine-7-sulfonamide 1,1-dioxide, 6-Chloro-3,4-dihydro-7-sulfamoyl-2H-1,2,4-benzothiadiazine 1,1-dioxide, 6-Chloro-7-sulfamoyl-3,4-dihydro-2H-1,2,4-benzothiadiazine 1,1-dioxide, 6-Chloro-7-sulfamoyl-3,4-dihydrobenzo-1,2,4-thiadiazine-1,1-dioxide, Acuretic, Apo-Hydro, Apresazide, Aquarius, Aquazide, Aquazide H, Caplaril, Carozide, Chlorosulthiadil, Chlorsulthiadil, Clothia, Dichlothiazide, Dichlotiazid, Dichlotride, Dichlozid, Diclotride, Didralin, Dihydrochlorothiazide, Dihydrochlorothiazidum, Dihydrochlorurit, Dihydrochlorurite, Disalunil, Disothiazide, Diuret-P, Diurex, Diurizid, Drenol, Esidrex, Esidrix, HCT, HCTZ, HCZ, HH 25/25, HH 50/50, Hidrenox, Hidril, Hidrochlortiazid, Hidrosaluretil, Hidrotiazida, Hydrex, Hydrex-semi, Hydril, Hydro Par, Hydro-Aquil, Hydro-Diuril, Hydro-T, Hydrochlorothiazid, Hydrochlorthiazide, Hydrochlorzide, Hydrocot, Hydrodiuretic, Hydrosaluric, Hydrozide, Hypothiazid, Hypothiazide, Hypothiazide (C7H8ClN3O4S2), Idrodiuvis, Idrotiazide, Lopressor HCT, Lotensin HCT, Manschitt, Maschitt, Megadiuril, Microzide, NSC 53477, Natrimax, Nefrix, Nefrol, Neoflumen, Newtolide, Oretic, Pantemon, Servithiazid, Su 5879, Tandiur, Thiuretic, Timolide, Unipres, Urirex, Vaseretic, Vetidrex, Ziac, Zide
IUPAC Name 6-chloro-1,1-dioxo-3,4-dihydro-2H-1λ^{6},2,4-benzothiadiazine-7-sulfonamide
SMILES NS(=O)(=O)c1cc2c(NCNS2(=O)=O)cc1Cl

Specifications

Quantity 25 g
InChI Formula InChI=1S/C7H8ClN3O4S2/c8-4-1-5-7(2-6(4)16(9,12)13)17(14,15)11-3-10-5/h1-2,10-11H,3H2,(H2,9,12,13)
Formula Weight 296.9645
Percent Purity >95%
Chemical Name or Material Hydrochlorothiazide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.