missing translation for 'onlineSavingsMsg'
Learn More

Hydrocinnamic Acid, TRC

Product Code. 30405194
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30405194 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30405194 Supplier Toronto Research Chemicals Supplier No. TRCH7145951G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hydrocinnamic Acid
CAS 501-52-0
Purity Grade Notes HPLC
Molecular Formula C9 H10 O2
Quantity 1 g
Synonym Benzenepropanoic Acid, Hydrocinnamic acid (8CI), 3-Phenyl-n-propionic acid, 3-Phenylpropanoic acid, 3-Phenylpropionic acid, Benzenepropionic acid, Benzylacetic acid, Dihydrocinnamic acid, NSC 9272, β-Phenylpropanoic acid, β-Phenylpropionic acid, ω-Phenylpropanoic acid, Benzenepropanoic Acid, Hydrocinnamic acid (8CI), 3-Phenyl-n-propionic acid, 3-Phenylpropanoic acid, 3-Phenylpropionic acid, Benzenepropionic acid, Benzylacetic acid, Dihydrocinnamic acid, NSC 9272, beta-Phenylpropanoic acid, beta-Phenylpropionic acid, omega-Phenylpropanoic acid
InChI Formula InChI=1S/C9H10O2/c10-9(11)7-6-8-4-2-1-3-5-8/h1-5H,6-7H2,(H,10,11)
SMILES OC(=O)CCc1ccccc1
IUPAC Name 3-phenylpropanoic acid
Molecular Weight (g/mol) 150.17
Formula Weight 150.0681
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.