missing translation for 'onlineSavingsMsg'
Learn More

Hydrocortisone 21-Acetate, TRC

Product Code. 18033920
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18033920 1 g 1g
18002020 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18033920 Supplier Toronto Research Chemicals Supplier No. TRCH7146201G
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-03-3
Molecular Formula C23 H32 O6
Molecular Weight (g/mol) 404.5
Synonym Hydrocortisone Acetate, Hydrocortisone 21-Acetate, Hydrocortisone Hydrogen Succinate Imp. B (EP), Prednisolone Acetate Imp. A (EP), Cortisone Acetate Imp. A (EP), Hydrocortisone Imp. C (EP), (11β)-21-(Acetyloxy)-11,17-dihydroxypregn-4-ene-3,20-dione, Cortisol acetate, Cortisol 21-acetate, 11β,17α,21-Trihydroxypregn-4-ene-3,20-dione 21-acetate, 17-Hydroxycorticosterone 21-acetate, 17α-Hydroxycorticosterone acetate, 21-Acetoxy-11β,17-dihydroxypregn-4-en-3,20-dione, 21-Acetoxy-11β,17-dihydroxypregn-4-ene-3,20-dione, 21-Acetoxy-11β,17α-dihydroxypregn-4-ene-3,20-dione, Abbocort, Anusol HC, Bambicort, Berlison F, Biocortar, CaldeCort, Chemysone, Colifoam, Colofoam, Cordes, Cortacream, Cortaid, Cortef acetate, Cortell, Corticaine, Cortifoam, Cortril acetate, Cortril acetate AS, Efcolin, HA, Hc 45, Hycortole acetate, Hydrin 2, Hydrocal, Hydrocort acetate, Hydrocortisat, Hydrocortisone 21-acetate, Hydrocortisone acetate, Hydrocortistab, Hydrocortone acetate, Hydroxycorticosterone acetate, Isopto-Hydrocortisone, Lanacort, Lenirit, NSC 741, Pabracort, Proctocort, Sigmacort, Sintotrat, Velopural, Wycort
IUPAC Name [2-[(8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-10,13-dimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
SMILES CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3[C@@H](O)C[C@]12C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C23H32O6/c1-13(24)29-12-19(27)23(28)9-7-17-16-5-4-14-10-15(25)6-8-21(14,2)20(16)18(26)11-22(17,23)3/h10,16-18,20,26,28H,4-9,11-12H2,1-3H3/t16-,17-,18-,20+,21-,22-,23-/m0/s1
Formula Weight 404.2199
Chemical Name or Material Hydrocortisone Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.