missing translation for 'onlineSavingsMsg'
Learn More

Hydrocortisone, 98%

Product Code. 11487747
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11487747 5 g 5g
11477747 1 g 1g
11497747 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 11487747 Fournisseur Thermo Scientific Alfa Aesar Code fournisseur A16292.06
Black Friday Starts Here
Use promo code 27984
Afficher toutes les promotions
 Restricted Item
In Stock
Ajouter au panier
Edge
Ajouter au panier
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 50-23-7 | C21H30O5 | 362.466 g/mol

Cortisol, steroid hormone is released in response to stress and a low level of blood glucocorticoids. Its primary functions is to increase blood sugar through gluconeogenesis, suppress the immune system, and aid in fat, protein and carbohydrate metabolism.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cortisol, steroid hormone is released in response to stress and a low level of blood glucocorticoids. Its primary functions is to increase blood sugar through gluconeogenesis, suppress the immune system, and aid in fat, protein and carbohydrate metabolism.

Solubility
It is soluble in DMSO and ethanol.

Notes
Protect against light. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 50-23-7
Molecular Formula C21H30O5
MDL Number MFCD00011654
InChI Key JYGXADMDTFJGBT-VWUMJDOOSA-N
Synonym hydrocortisone, cortisol, acticort, cetacort, cortef, 17-hydroxycorticosterone, hydrocortisyl, hydrasson, cobadex, cortril
PubChem CID 5754
ChEBI CHEBI:17650

Spécification

Melting Point 213°C to 215°C
Assay Percent Range 98%
Quantity 5 g
Beilstein 1354819
Merck Index 14,4787
Solubility Information It is soluble in DMSO and ethanol.
SMILES CC12CCC(=O)C=C1CCC3C2C(CC4(C3CCC4(C(=O)CO)O)C)O
IUPAC Name (8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one
Molecular Weight (g/mol) 362.466
Formula Weight 362.47
Percent Purity 98%
Chemical Name or Material Hydrocortisone
Afficher plus Afficher moins
compliance-icons
Identifiant produit
  • Hydrocortisone
Mention d'avertissement
  • Danger
Catégorie de danger
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
Mention de danger
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
Conseils de prudence
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Informations supplémentaires
  • MIXTURE LIST-Contain: Hydrocortisone

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.