missing translation for 'onlineSavingsMsg'
Learn More

Hydrocortisone, 98%, Thermo Scientific™

35.30 € - 120.00 €

Chemical Identifiers

CAS 50-23-7
Molecular Formula C21H30O5
Molecular Weight (g/mol) 362.466
MDL Number MFCD00011654
InChI Key JYGXADMDTFJGBT-VWUMJDOOSA-N
Synonym hydrocortisone, cortisol, acticort, cetacort, cortef, 17-hydroxycorticosterone, hydrocortisyl, hydrasson, cobadex, cortril
PubChem CID 5754
ChEBI CHEBI:17650
IUPAC Name (8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one
SMILES CC12CCC(=O)C=C1CCC3C2C(CC4(C3CCC4(C(=O)CO)O)C)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11477747
SDS View Documents
Thermo Scientific Alfa Aesar
A16292.03
1g
35.30 €
1g
View alternative products
Save Up to 
»
In Stock
Edge
Add to basket
 
 Restricted Item
11487747
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
A16292.06
5g
120.00 €
5g
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Cortisol, steroid hormone is released in response to stress and a low level of blood glucocorticoids. Its primary functions is to increase blood sugar through gluconeogenesis, suppress the immune system, and aid in fat, protein and carbohydrate metabolism.

Solubility
It is soluble in DMSO and ethanol.

Notes
Protect against light. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

50-23-7
362.466
JYGXADMDTFJGBT-VWUMJDOOSA-N
5754
(8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one
C21H30O5
MFCD00011654
hydrocortisone, cortisol, acticort, cetacort, cortef, 17-hydroxycorticosterone, hydrocortisyl, hydrasson, cobadex, cortril
CHEBI:17650
CC12CCC(=O)C=C1CCC3C2C(CC4(C3CCC4(C(=O)CO)O)C)O

Specifications

50-23-7
Odorless
MFCD00011654
14,4787
It is soluble in DMSO and ethanol.
CC12CCC(=O)C=C1CCC3C2C(CC4(C3CCC4(C(=O)CO)O)C)O
362.466
CHEBI:17650
98%
98%
C21H30O5
1354819
hydrocortisone, cortisol, acticort, cetacort, cortef, 17-hydroxycorticosterone, hydrocortisyl, hydrasson, cobadex, cortril
JYGXADMDTFJGBT-VWUMJDOOSA-N
(8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one
5754
362.47
Hydrocortisone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H361
Suspected of damaging fertility or the unborn child.

P201-P202-P260-P281-P308+P313-P501c

H360Df-H373

missing translation for 'einecsNumber' : 200-020-1

missing translation for 'rtecsNumber' : GM8925000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Hydrocortisone

Signal Word
  • Danger

Hazard Category
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contain: Hydrocortisone

SDS
Documents

Documents

Certificates

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.