missing translation for 'onlineSavingsMsg'
Learn More

Hydrocortisone (9Beta,11Beta)-Epoxide, TRC

Product Code. 17954345
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17954345 50 mg 50mg
17904745 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17954345 Supplier Toronto Research Chemicals Supplier No. TRCH71459050MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10072-97-6
Molecular Formula C21 H28 O5
Molecular Weight (g/mol) 360.44
Synonym Pregn-4-ene-3,20-dione, 9,11-epoxy-17,21-dihydroxy-, (9β,11β)- (9CI), 9β-Pregn-4-ene-3,20-dione, 9,11β-epoxy-17,21-dihydroxy- (6CI,7CI,8CI), 9,11-Epoxy-9H-cyclopenta[a]phenanthrene, pregn-4-ene-3,20-dione deriv., 9β,11β-Epoxy-17,21-dihydroxypregn-4-ene-3,20-dione
IUPAC Name (1S,2S,10S,11S,14R,15S,17S)-14-hydroxy-14-(2-hydroxyacetyl)-2,15-dimethyl-18-oxapentacyclo[8.8.0.01,17.02,7.011,15]octadec-6-en-5-one
SMILES C[C@]12C[C@@H]3O[C@]34[C@@H](CCC5=CC(=O)CC[C@]45C)[C@@H]1CC[C@]2(O)C(=O)CO

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C21H28O5/c1-18-7-5-13(23)9-12(18)3-4-15-14-6-8-20(25,16(24)11-22)19(14,2)10-17-21(15,18)26-17/h9,14-15,17,22,25H,3-8,10-11H2,1-2H3/t14-,15-,17-,18-,19-,20-,21+/m0/s1
Formula Weight 360.1937
Percent Purity >95%
Chemical Name or Material Hydrocortisone (9beta,11beta)-Epoxide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.