missing translation for 'onlineSavingsMsg'
Learn More

Hydrocortisone acetate, 97+%

Product Code. 11439236
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11439236 1 g 1g
11449236 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11439236 Supplier Thermo Scientific Alfa Aesar Supplier No. A18089.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 50-03-3 | C23H32O6 | 404.50 g/mol

Hydrocortisone acetate is used to treat hemorrhoids and itching/swelling in the rectum and anus. It is used to treat certain intestinal problems (such as ulcerative colitis of the rectum and other rectal/anal inflammatory conditions).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store in the dark.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-03-3
Molecular Formula C23H32O6
Molecular Weight (g/mol) 404.50
MDL Number MFCD00037714
InChI Key ALEXXDVDDISNDU-JZYPGELDSA-N
Synonym hydrocortisone acetate, cortell, cortisol 21-acetate, cortisol acetate, cortacream, hydrocortisat, abbocort, bambicort, biocortar, chemysone
PubChem CID 5744
ChEBI CHEBI:17609
SMILES CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3[C@@H](O)C[C@]12C

Specifications

Melting Point ∼220°C (decomposition)
Quantity 1 g
Beilstein 2066841
Sensitivity Light sensitive
Merck Index 14,4787
Solubility Information Insoluble in water.
IUPAC Name [2-[(8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-10,13-dimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
Formula Weight 404.51
Percent Purity ≥97%
Chemical Name or Material Hydrocortisone acetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.