missing translation for 'onlineSavingsMsg'
Learn More

Hydrocortisone (Cortisol), TRC

Product Code. 18092910
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18092910 1 g 1g
18021020 250 mg 250mg
18011020 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18092910 Supplier Toronto Research Chemicals Supplier No. TRCH7146151G
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-23-7
Molecular Formula C21 H30 O5
Molecular Weight (g/mol) 362.46
Synonym Hydrocortisone, Cortisol, 11beta,17,21-Trihydroxy-pregn-4-ene-3,20-dione, Hydrocortisone Hydrogen Succinate Imp. A (EP), Prednisolone Imp. A (EP), Hydrocortisone Acetate Imp. A (Pharmeuropa), 11β,17,21-Trihydroxyprogesterone, 11β,17α,21-Trihydroxypregn-4-ene-3,20-dione, 11β-Hydroxycortisone, 17-Hydroxycorticosterone, 17α-Hydroxycorticosterone, 4-Pregnene-11β,17α,21-triol-3,20-dione, Acticort, Aeroseb HC, Ala-Cort, Anflam, Anti-inflammatory hormone, CCN 90306A, CaldeCort Spray, Cetacort, Cobadex, Cort-Dome, Cortanal, Cortef, Cortenema, Corticreme, Cortifan, Cortiment, Cortispray, Cortonema, Cortril, Dermacort, Dermocortal, Dermolate, Dihydrocostisone, Dioderm, Domolene-HC, Efcorbin, Efcortelan, Eldecort, Epiderm H, Esiderm H, Evacort, Ficortril, Genacort, HC, Heb-Cort, Hidro-Colisona, Hycort, Hycortol, Hycortole, Hydracort, Hydrasson, Hydro-Adreson, Hydrocort, Hydrocortal, Hydrocorticosterone, Hydrocortisone, Hydrocortisyl, Hydrocortone, Hydroskin, Hytone, Hytone lotion, Incortin-H, Kendall's compound F, Lacticare HC, Medicort, Mildison, NSC 10483, Nutracort, Optef, Otosone-F, Penecort, Permicort, Polcort H, Reichstein's substance M, Scheroson F, Signef, Synacort, Tarcortin, Texacort, Texacort lotion 25, Timocort, Topicort, Traumaide, U 1851, Zenoxone
IUPAC Name (8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one
SMILES C[C@]12CCC(=O)C=C1CC[C@H]3[C@@H]4CC[C@](O)(C(=O)CO)[C@@]4(C)C[C@H](O)[C@H]23

Specifications

Quantity 1 g
InChI Formula InChI=1S/C21H30O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h9,14-16,18,22,24,26H,3-8,10-11H2,1-2H3/t14-,15-,16-,18+,19-,20-,21-/m0/s1
Formula Weight 362.2093
Percent Purity >95%
Chemical Name or Material Hydrocortisone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.