missing translation for 'onlineSavingsMsg'
Learn More

Hydroflumethiazide, TRC

Product Code. 30137370
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137370 250 mg 250mg
30004871 100 mg 100mg
30008183 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137370 Supplier Toronto Research Chemicals Supplier No. TRCH714630250MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135-09-1
Molecular Formula C8 H8 F3 N3 O4 S2
Molecular Weight (g/mol) 331.29
Synonym 2H-1,2,4-Benzothiadiazine-7-sulfonamide, 3,4-dihydro-6-(trifluoromethyl)-, 1,1-dioxide, 3,4-Dihydro-6-trifluoromethyl-2H-1,2,4-benzothiadiazine-7-sulfonamide 1,1-dioxide, 3,4-Dihydro-6-trifluoromethyl-2H-1,2,4-benzothiazide-7-sulfonamide 1,1-dioxide, 3,4-Dihydro-6-trifluoromethyl-7-sulfamoylbenzo-1,2,4-thiadiazine 1,1-dioxide, 3,4-Dihydro-7-sulfamoyl-6-trifluoromethyl-2H-1,2,4-benzothiadiazine 1,1-dioxide, 3,4-Dihydro-7-sulfamyl-6-trifluoromethyl-1,2,4-benzothiadiazine 1,1-dioxide, 3,4-Dihydro-7-sulfamyl-6-trifluoromethyl-2H-1,2,4-benzothiadiazine 1,1-dioxide, 6-Trifluoromethyl-3,4-dihydro-7-sulfamoyl-2H-1,2,4-benzothiadiazine 1,1-dioxide, 6-Trifluoromethyl-7-sulfamyl-3,4-dihydro-1,2,4-benzothiadiazine-1,1-dioxide, 7-Sulfamyl-6-trifluoromethyl-3,4-dihydro-1,2,4-benzothiadiazine 1,1-dioxide, Bristab, Bristurin, Di-Ademil, Dihydroflumethiazide, Diucardin, Diumide K, Diuredemina, Diurometon, Elodrine, Finuret, Flutizide, Glomerulin, Hidroalogen, Hidroflumetiazid, Hydol, Hydrenox, Hydroflumethiazide, Leodrine, Metflorylthiazidine, Methforylthiazidine, NSC 44627, NaClex, NaClex (Glaxo), Olmagran, Rodiuran, Rontyl, Saluron, Salutensin, Sisuril, Spandiuril, Trifluoromethylhydrothiazide, Vergonil
IUPAC Name 1,1-dioxo-6-(trifluoromethyl)-3,4-dihydro-2H-1λ^{6},2,4-benzothiadiazine-7-sulfonamide
SMILES NS(=O)(=O)c1cc2c(NCNS2(=O)=O)cc1C(F)(F)F

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C8H8F3N3O4S2/c9-8(10,11)4-1-5-7(2-6(4)19(12,15)16)20(17,18)14-3-13-5/h1-2,13-14H,3H2,(H2,12,15,16)
Formula Weight 330.9908
Percent Purity >95%
Chemical Name or Material Hydroflumethiazide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.