Product Code. 15680530
Shop All Honeywell Fluka Products
Click to view available options
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
unitSize
15680530
5 L
5 L
15670530
30 kg
30kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code.
15680530
Supplier
Honeywell-Fluka
Supplier No.
010105L
This item is currently unavailable or has been discontinued. View the product page for possible alternatives.
View alternative products
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Puriss.,≥40%
Leading the Industry For Over 65 Years Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemische Identifikatoren
CAS
7664-39-3
Molecular Formula
FH
Molecular Weight (g/mol)
20.01
MDL Number
MFCD00011346
InChI Key
KRHYYFGTRYWZRS-UHFFFAOYSA-N
Synonym
hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484
PubChem CID
14917
ChEBI
CHEBI:29228
IUPAC Name
fluorane
SMILES
F
Spezifikation
Concentration
≥40%
Density
1.15g/mL at 25°C
Vapor Pressure
25mmHg (20°C)
Quantity
5 L
UN Number
1790
Formula Weight
20.01g/mol
Grade
puriss.
Chemical Name or Material
Hydrofluoric Acid
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt.
Datenschutzrichtlinie .
Haben Sie Verbesserungsvorschläge?