missing translation for 'onlineSavingsMsg'
Learn More

Hydrofluoric acid, Puriss. p.a., Reag. ISO, Reag. Ph. Eur., ≥40%, Solstice

Product Code. 15642980
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
Unit Size:
1 L
2.5 L
Pack of 4
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15642980 6 x 1 L Pack of 6
15602980 1 L 1 L
15612980 2.5 L 2.5 L
15622980 4 x 2.5 L Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15642980 Supplier Honeywell-Fluka Supplier No. 301036X1L
  • ONLINE EXCLUSIVE PRICE
    740.00 €
    167.90 € / Pack of 6
    Save 572.10 €
    77% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss. p.a., Reag. ISO, Reag. Ph. Eur.,≥40%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-39-3 , 7732-18-5
Molecular Formula FH
Molecular Weight (g/mol) 20.01
MDL Number MFCD00011346
InChI Key KRHYYFGTRYWZRS-UHFFFAOYSA-N
Synonym hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484
PubChem CID 14917
ChEBI CHEBI:29228

Specifications

Boiling Point 110°C
CAS Min % ≥50.0000%, ≥30.0000%
CAS Max % <70.0000%, <50.0000%
Density 1.15 g/mL at 25 °C(lit.)
Packaging Poly Bottle
Quantity 6 x 1 L
Linear Formula HF
UN Number UN1790
SMILES [H+].[F-]
IUPAC Name fluorane
Formula Weight 20.01
Percent Purity >40%
Grade puriss. p.a.
Chemical Name or Material Hydrofluoric acid, Puriss. p.a., Reag. ISO, Reag. Ph. Eur., ≥40%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Hydrofluoric acid
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 2
  • Acute toxicity Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H300+H310+H330-Fatal if swallowed, in contact with skin or if inhaled
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Hydrogen fluoride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.