missing translation for 'onlineSavingsMsg'
Learn More

Hydrofluoric Acid, Technical grade, Solstice

Product Code. 15610540
Change view
Click to view available options
Quantity:
1 L
5 L
6 x 1 L
Unit Size:
1 L
5 L
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15610540 6 x 1 L Pack of 6
15690530 1 L 1 L
15600540 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15610540 Supplier Honeywell-Fluka Supplier No. 010666X1L
  • ONLINE EXCLUSIVE PRICE
    282.00 €
    264.64 € / Pack of 6
    Save 17.36 €
    6% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Technical, 40-45%

  • Chemicals that do not have an established standard set for quality and impurity levels or for products where the purity is <90%
  • Variation in color and physical form is possible
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-39-3
Molecular Formula FH
Molecular Weight (g/mol) 20.01
MDL Number MFCD00011346
InChI Key KRHYYFGTRYWZRS-UHFFFAOYSA-N
Synonym hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484
PubChem CID 14917
ChEBI CHEBI:29228

Specifications

Density 1.15g/mL at 25°C
Vapor Pressure 25mmHg (20°C)
Quantity 6 x 1 L
UN Number UN1790
SMILES [H+].[F-]
IUPAC Name hydrogen fluoride
Formula Weight 20.01
Percent Purity 40 to 45%
Grade Technical
Chemical Name or Material Hydrofluoric Acid
compliance-icons compliance-icons
Product Identifier
  • Hydrofluoric acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Hydrogen fluoride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.