missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrofluoric Acid, TRC
Specifications
Specifications
| Chemical Name or Material | Hydrofluoric Acid |
| CAS | 7664-39-3 |
| Quantity | 10 g |
| Molecular Formula | FH |
| Synonym | Alsurf 45, Anhydrous hydrofluoric Acid, Antisal 2b, Fluorhydric Acid, Fluoric Acid, Fluorine Hydride (FH), Fluorine Monohydride, G-C Hydrofluoride Gel, Hydrofluoric Acid Gas, Hydrogen Fluoride, Hydrogen Fluoride (HF), Hydrogen Monofluoride, Porcelain Conditioner, SA-X, Ultradent Porcelain Etch, Urine Luck 6.3, Vita Ceramic etch |
| InChI Formula | InChI=1S/FH/h1H |
| SMILES | F |
| IUPAC Name | fluorane |
| Molecular Weight (g/mol) | 20.0064 |
| Formula Weight | 20.0062 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?