missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen bromide, pure, 33 wt% solution in glacial acetic acid

Product Code. 11334996 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11334996 2.5 L Glass bottle 2.5 L
10011200 500 mL Glass bottle 500mL
10626132 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11334996 Supplier Thermo Scientific Acros Supplier No. 123180025
  • ONLINE EXCLUSIVE PRICE
    277.00 €
    255.00 € / 2.5 L
    Save 22.00 €
    7% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Hydrogen bromide, 31 to 34 wt% (HBr), BrH, CAS Number-10035-10-6, 64-19-7, acide bromhydrique, acido bromidrico, anhydrous hydrobromic acid, bromowodor, bromowodor polish, bromwasserstoff, broomwaterstof, hydrobromic acid, hydrobromide, hydrogen bromide, 2.5L, Vapor Pressure: 300mmHg at 20 deg.C | CAS: 10035-10-6 | BrH | 80.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10035-10-6 , 64-19-7
Molecular Formula BrH
Molecular Weight (g/mol) 80.91
MDL Number MFCD00011323
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-N
Synonym hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
PubChem CID 260
ChEBI CHEBI:47266
SMILES Br

Specifications

Color Yellow
Physical Form Fuming Liquid
Quantity 2.5 L
Linear Formula HBr
UN Number 2922
Fieser 01,450; 02,214; 03,154; 04,249; 05,332; 06,282; 10,200; 15,273
Flash Point 39°C
Merck Index 09, 4665
IUPAC Name hydrogen bromide
Formula Weight 80.9
Percent Purity 31 to 34 wt% (HBr)
Grade Pure
Packaging Glass bottle
Specific Gravity 1.42
Vapor Pressure 300mmHg at 20°C
Density 1.42 g/mL
Chemical Name or Material Hydrogen bromide, Pure
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hydrogen bromide, 33 wt% solution in glacial acetic acid
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Acetic acid, Hydrogen bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.