missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen fluoride-pyridine, approx. 70% HF, approx. 30% Pyridine

Product Code. 10502401 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Packaging:
Plastic Bottle in Tin Can
Plastic bottle
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10502401 100 g Plastic Bottle in Tin Can 100g
10317220 10 g Plastic bottle 10g
10773371 50 g Plastic Bottle in Tin Can 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10502401 Supplier Thermo Scientific Supplier No. 180071000
  • ONLINE EXCLUSIVE PRICE
    119.00 €
    111.00 € / 100g
    Save 8.00 €
    6% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 32001-55-1 | C5H6FN | 99.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32001-55-1
Molecular Formula C5H6FN
Molecular Weight (g/mol) 99.11
MDL Number MFCD00012436
InChI Key GRJJQCWNZGRKAU-UHFFFAOYSA-N
Synonym pyridine hydrofluoride, hydrogen fluoride-pyridine, olah's reagent, pyridinium fluoride, hydrogen fluoride pyridine complex, hydrogen fluoride pyridine, hf-pyridine, pyridine, hydrofluoride 1:1, pyridinium poly hydrogen fluoride, hydrogen fluoride/pyridine
PubChem CID 64774
IUPAC Name pyridine;hydrofluoride
SMILES C1=CC=NC=C1.F

Specifications

Color Yellow to Brown
Density 1.1000g/mL
Assay Percent Range 65-70%
Packaging Plastic Bottle in Tin Can
Quantity 100 g
Fieser 16,286; 16,050; 05,538; 06,473; 09,399; 12,419
Specific Gravity 1.1
Formula Weight 99.11
Percent Purity 65-70%
Physical Form Liquid
Chemical Name or Material Hydrogen fluoride-pyridine, 65-70%
Show More Show Less
compliance-icons
Product Identifier
  • Hydrogen fluoride-pyridine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.