missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen fluoride-pyridine, approx. 70% HF, approx. 30% Pyridine

Product Code. 10773371 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
100 g
Packaging:
Plastic Bottle in Tin Can
Plastic bottle
Unit Size:
100g
10g
50g
This item is not returnable. View return policy

Product Code. 10773371

Brand: Thermo Scientific Acros 180070500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 32001-55-1 | C5H6FN | 99.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32001-55-1
Molecular Formula C5H6FN
Molecular Weight (g/mol) 99.11
MDL Number MFCD00012436
InChI Key GRJJQCWNZGRKAU-UHFFFAOYSA-N
Synonym pyridine hydrofluoride, hydrogen fluoride-pyridine, olah's reagent, pyridinium fluoride, hydrogen fluoride pyridine complex, hydrogen fluoride pyridine, hf-pyridine, pyridine, hydrofluoride 1:1, pyridinium poly hydrogen fluoride, hydrogen fluoride/pyridine
PubChem CID 64774
IUPAC Name pyridine;hydrofluoride
SMILES C1=CC=NC=C1.F

Specifications

Color Yellow to Brown
Density 1.1000g/mL
Assay Percent Range 65-70%
Packaging Plastic Bottle in Tin Can
Quantity 50 g
Fieser 16,286; 16,050; 05,538; 06,473; 09,399; 12,419
Specific Gravity 1.1
Formula Weight 99.11
Percent Purity 65-70%
Physical Form Liquid
Chemical Name or Material Hydrogen fluoride-pyridine, 65-70%
Show More Show Less
compliance-icons
Product Identifier
  • Hydrogen fluoride-pyridine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.