missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen fluoride pyridine complex, ca 70% HF, Thermo Scientific Chemicals

Product Code. 11373956
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11373956 25 g 25g
11383956 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11373956 Supplier Thermo Scientific Alfa Aesar Supplier No. L17117.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hydrogen fluoride pyridine complex is used to prepare beta-fluoroamines via reaction with amino alcohols. As a fluorinating agent, is involved in the fluorination of 2,2-diary-1,3-dithiolanes. It is associated with nitrosonium tetrafluorborate to prepare gem-difluorides by reacting with ketoximes. It is also used to convert secondary and tertiary alcohols into their respective alkyl fluorides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hydrogen fluoride pyridine complex is used to prepare beta-fluoroamines via reaction with amino alcohols. As a fluorinating agent, is involved in the fluorination of 2,2-diary-1,3-dithiolanes. It is associated with nitrosonium tetrafluorborate to prepare gem-difluorides by reacting with ketoximes. It is also used to convert secondary and tertiary alcohols into their respective alkyl fluorides.

Notes
Moisture sensitive. Store in cool place. Incompatible with strong bases, alkali metals, strong oxidizing agents, metals and strong acids. It reacts violently with water.
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 62778-11-4
Molecular Formula C5H6FN
Molecular Weight (g/mol) 99.108
MDL Number MFCD00012436
InChI Key GRJJQCWNZGRKAU-UHFFFAOYSA-N
Synonym pyridine hydrofluoride, hydrogen fluoride-pyridine, olah's reagent, pyridinium fluoride, hydrogen fluoride pyridine complex, hydrogen fluoride pyridine, hf-pyridine, pyridine, hydrofluoride 1:1, pyridinium poly hydrogen fluoride, hydrogen fluoride/pyridine
PubChem CID 64774
IUPAC Name pyridine;hydrofluoride
SMILES C1=CC=NC=C1.F

Tekniske data

Density 1.1
Boiling Point 50°C (1 mmHg)
Quantity 25 g
UN Number UN1790
Beilstein 4750151
Sensitivity Moisture sensitive
Merck Index 14,6821
Percent Purity ca. 70% HF
Physical Form Liquid
Assay ≈70% HF
Chemical Name or Material Hydrogen fluoride pyridine complex
Vis mere Vis mindre
compliance-icons compliance-icons
Produktidentifikator
  • Hydrogen fluoride pyridine complex
Signalord
  • Danger
Farekategori
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Faresætning
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
Sikkerhedssætning
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
Supplerende oplysninger
  • MIXTURE LIST-Contain: Hydrogen fluoride pyridine

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.